3,3'-Difluorbiphenyl ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 3,3'-Difluoro-1,1'-biphenyl

L16740.03 L16740.06
L16740.03EA 92.2 EUR
L16740.03 L16740.06
3,3'-Difluorbiphenyl ≥97%
3,3'-Difluorbiphenyl

Purity: 97% CAS No.: 396-64-5 Molecular Formula: C12H8F2 Formula Weight: 190.19 Refractive Index: 1.5680 Melting Point: 7-8° Boiling Point: 130°/14mm Beilstein Reference: 2046445 UN No.: 3151 MDL No.: MFCD00039216

Formel: C₁₂H₈F₂
Molecular Weight: 190.19 g/mol
Siedepunkt: 130 °C (14 mmHg)
Schmelzpunkt: 7…8 °C
Dichte: 1.210
Lagertemperatur: Raumtemperatur
MDL: MFCD00039216
CAS-Nummer: 396-64-5
EINECS: 206-905-9
ADR: 9 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR