3-Fluor-L-tyrosin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Fluoro-4-hydroxy-L-phenylalanine, (S)-2-Amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid, H-Tyr(3-F)-OH
L16775.MF
L16775.MD
L16775.MFEA
60.1
EUR
L16775.MF
L16775.MD
3-Fluor-L-tyrosin ≥97%
3-Fluor-L-tyrosin
Purity: 97%
CAS No.: 7423-96-3
Molecular Formula: C9H10FNO3
Formula Weight: 199.18
Melting Point: 260-261°
Beilstein Reference: 3204802
UN No.: 2811
MDL No.: MFCD00002607
Formel:
C₉H₁₀FNO₃ Schmelzpunkt: 260…261 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002607 CAS-Nummer: 7423-96-3 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...