1H,1H,10H,10H-Hexadecafluor-1,10-decandiol 96%
Lieferant: Thermo Scientific
Synonyme:
1H,1H,10H,10H-Perfluor-1,10-decandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluor-1,10-decandiol
L16823.06
L16823.06EA
70.8
EUR
L16823.06
1H,1H,10H,10H-Hexadecafluor-1,10-decandiol 96%
1H,1H,10H,10H-Hexadecafluor-1,10-decandiol
Purity: 96%
CAS No.: 754-96-1
Molecular Formula: HOCH2(CF2)8CH2OH
Formula Weight: 462.13
Melting Point: 132-134°
Boiling Point: 132°/4mm
MDL No.: MFCD00192200
Formel:
C₁₀H₆F₁₆O₂ Molecular Weight: 462,13 g/mol Siedepunkt: 132 °C (4 mmHg) Schmelzpunkt: 132…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192200 CAS-Nummer: 754-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...