4,4,4-Trifluorocrotononitrile 96%

Lieferant: Thermo Scientific
Gefahr

L16870.03
L16870.03EA 89 EUR
L16870.03
4,4,4-Trifluorocrotononitrile 96%
4,4,4-Trifluorocrotononitrile

Purity: 96% CAS No.: 406-86-0 Molecular Formula: CF3CH=CHCN Formula Weight: 121.06 Boiling Point: 84-85° Beilstein Reference: 6381034 UN No.: UN3276 MDL No.: MFCD00190645

Formel: CF₃CH=CHCN
Molecular Weight: 121,06 g/mol
Siedepunkt: 84…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00190645
CAS-Nummer: 406-86-0
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR