3,3,3-Trifluor-1-propanol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,3,3-Trifluoropropan-1-ol

L16879.03 L16879.06
L16879.03EA 87 EUR
L16879.03 L16879.06
3,3,3-Trifluor-1-propanol ≥97%
3,3,3-Trifluor-1-propanol

Purity: 97% CAS No.: 2240-88-2 Molecular Formula: CF3CH2CH2OH Formula Weight: 114.07 Flash Point: 37°(98°F) Refractive Index: 1.3200 Boiling Point: 100-101° Beilstein Reference: 1734777 UN No.: 1987 MDL No.: MFCD00190649

Formel: C₃H₅F₃O
Molecular Weight: 114.07 g/mol
Siedepunkt: 100…101 °C
Dichte: 1.294
Flammpunkt: 37 °C (98 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00190649
CAS-Nummer: 2240-88-2
UN: 1987
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR