2-tert-Butylpropane-1,3-diol 98%
Lieferant: Thermo Fisher Scientific
L16927.06
L16927.06EA
158
EUR
L16927.06
2-tert-Butylpropane-1,3-diol 98%
2-tert-Butylpropane-1,3-diol
Purity: 98%
CAS No.: 2819-05-8
Molecular Formula: C7H16O2
Formula Weight: 132.20
Melting Point: 54-56°
Boiling Point: 100-102°/2mm
Beilstein Reference: 1900687
MDL No.: MFCD01075745
Formel:
C₇H₁₆O₂ Siedepunkt: 100…102 °C (2 mmHg) Schmelzpunkt: 54…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075745 CAS-Nummer: 2819-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...