2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-[N,N-Bis(Trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)triflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonyl)imide, Comins Triflating Reagent, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonimide)
L16929.03
L16929.06
L16929.03EA
50.4
EUR
L16929.03
L16929.06
2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine 99%
2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine
Purity: 99%
CAS No.: 145100-51-2
Molecular Formula: C7H3ClF6N2O4S2
Formula Weight: 392.68
Storage Temperature: 2°C to 8°C
Melting Point: 44-47°
Beilstein Reference: 5833971
MDL No.: MFCD00191833
Formel:
C₇H₃ClF₆N₂O₄S₂ Molecular Weight: 392.67 g/mol Schmelzpunkt: 44…47 °C |
MDL:
MFCD00191833 CAS-Nummer: 145100-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...