2H-Perfluoro-5-methyl-3,6-dioxanonane 97%

Lieferant: Thermo Fisher Scientific

L16954.06
L16954.06EA 119 EUR
L16954.06
2H-Perfluoro-5-methyl-3,6-dioxanonane 97%
2H-Perfluoro-5-methyl-3,6-dioxanonane

Purity: 97% CAS No.: 3330-14-1 Molecular Formula: C8HF17O2 Formula Weight: 452.07 Flash Point: None Refractive Index: 1.3005 Melting Point: -122° Boiling Point: 103-105° MDL No.: MFCD00054714

Formel: C₈HF₁₇O₂
Siedepunkt: 103…105 °C
Schmelzpunkt: –122 °C
Dichte: 1,659
Lagertemperatur: Raumtemperatur
MDL: MFCD00054714
CAS-Nummer: 3330-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR