ω-Chlor-ω,ω-difluoracetophenon 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Chlor-2,2-difluoracetophenon, α-Chlor-α,α-difluoracetophenon
L16963.06
AB180285.0005
L16963.06EA
99.6
EUR
L16963.06
ABCRAB180285.0005
ω-Chlor-ω,ω-difluoracetophenon 96%
ω-Chlor-ω,ω-difluoracetophenon
Purity: 96%
CAS No.: 384-67-8
Molecular Formula: C8H5ClF2O
Formula Weight: 190.58
Flash Point: 72°(161°F)
Refractive Index: 1.4970
Boiling Point: 95-97°/37mm
Beilstein Reference: 2251448
MDL No.: MFCD00042123
Formel:
(C₆H₅)C(O)CClF₂ Siedepunkt: 95…97 °C (37 mmHg) Dichte: 1,293 Flammpunkt: 72 °C (161 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042123 CAS-Nummer: 384-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...