Decyltriethoxysilan ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Triethoxy(decyl)silane

L16965.14 L16965.22
L16965.14EA 91.3 EUR
L16965.14 L16965.22
Decyltriethoxysilan ≥98%
Decyltriethoxysilan

Purity: 98% CAS No.: 2943-73-9 Molecular Formula: C16H36O3Si Formula Weight: 304.55 Flash Point: 110°(230°F) Refractive Index: 1.4220 Boiling Point: 150°/8mm Beilstein Reference: 2413155 MDL No.: MFCD00069132

Formel: C₁₆H₃₆O₃Si
Molecular Weight: 304.55 g/mol
Siedepunkt: 150 °C (8 mmHg)
Dichte: 0.879
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00069132
CAS-Nummer: 2943-73-9
EINECS: 220-940-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR