7-Isochinolinol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 7-Hydroxyisoquinoline, Isoquinolin-7-ol

L17067.MD
L17067.MDEA 51.7 EUR
L17067.MD
7-Isochinolinol ≥97%
7-Isochinolinol

Purity: 97% CAS No.: 7651-83-4 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 226-228° Beilstein Reference: 113224

Formel: C₉H₇NO
Molecular Weight: 145.16 g/mol
Schmelzpunkt: 226…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00456131
CAS-Nummer: 7651-83-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR