5-Methyl-1H-indole-2-carboxylic acid 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L17145.03
L17145.06
L17145.03EA
71.1
EUR
L17145.03
L17145.06
5-Methyl-1H-indole-2-carboxylic acid 99%
5-Methyl-1H-indole-2-carboxylic acid
Purity: 99%
CAS No.: 10241-97-1
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Melting Point: ca 235° dec.
Beilstein Reference: 144055
MDL No.: MFCD00047166
Formel:
C₁₀H₉NO₂ Schmelzpunkt: ∼ 235 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047166 CAS-Nummer: 10241-97-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...