D-Proline methyl ester hydrochloride 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
H-D-Pro-OMe.HCl
L17259.03
L17259.03EA
39.6
EUR
L17259.03
D-Proline methyl ester hydrochloride 99%
D-Proline methyl ester hydrochloride
Purity: 99%
CAS No.: 65365-28-8
Molecular Formula: C6H11NO2.HCl
Formula Weight: 165.62
Storage Temperature: 2°C to 8°C
Melting Point: 69-71°
Beilstein Reference: 4714125
Optical Rotation: +31° (c=1 in water)
MDL No.: MFCD00083685
Formel:
C₆H₁₁NO₂ Molecular Weight: 165.62 g/mol Schmelzpunkt: 69…71 °C |
MDL:
MFCD00083685 CAS-Nummer: 65365-28-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...