1-Chloro-3-fluoro-2-propanol 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L17286.06
L17286.06EA
146
EUR
L17286.06
1-Chloro-3-fluoro-2-propanol 95%
1-Chloro-3-fluoro-2-propanol
Purity: 95%
CAS No.: 453-11-2
Molecular Formula: C3H6ClFO
Formula Weight: 112.53
Refractive Index: 1.4280
Boiling Point: 153-156°
Beilstein Reference: 1732501
UN No.: UN1992
MDL No.: MFCD00045284
Formel:
C₃H₆ClFO Siedepunkt: 153…156 °C Dichte: 1,300 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045284 CAS-Nummer: 453-11-2 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...