3-Cyan-5-methylpyridin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Methylnicotinonitril, 3-Cyan-5-picolin, 5-Methyl-3-pyridincarbonitril

L17427.03 L17427.MD
L17427.03EA 77.5 EUR
L17427.03 L17427.MD
3-Cyan-5-methylpyridin ≥98%
3-Cyano-5-methylpyridine

Purity: 98+% CAS No.: 42885-14-3 Molecular Formula: C7H6N2 Formula Weight: 118.14 Melting Point: 82-85° Beilstein Reference: 111258

Formel: C₇H₆N₂
Molecular Weight: 118.14 g/mol
Siedepunkt: 220
Schmelzpunkt: 82…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00673151
CAS-Nummer: 42885-14-3
EINECS: 255-988-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR