Diphenyliodoniumtrifluormethansulfonat ≥99%

Lieferant: Thermo Scientific
Gefahr

L17444.03
L17444.03EA 74.5 EUR
L17444.03
Diphenyliodoniumtrifluormethansulfonat ≥99%
Diphenyliodonium trifluoromethanesulphonate

Purity: 99% CAS No.: 66003-76-7 Molecular Formula: C13H10F3IO3S Formula Weight: 430.19 Melting Point: 179-182° Beilstein Reference: 3582403 UN No.: 2811 MDL No.: MFCD00191356

Formel: C₁₃H₁₀F₃IO₃S
Molecular Weight: 430,19 g/mol
Schmelzpunkt: 179…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191356
CAS-Nummer: 66003-76-7
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR