5-Brom-2-iodpyrimidin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L17453.06
L17453.06EA 142 EUR
L17453.06
5-Brom-2-iodpyrimidin ≥98%
5-Bromo-2-iodopyrimidine

Purity: 98% CAS No.: 183438-24-6 Molecular Formula: C4H2BrIN2 Formula Weight: 284.88 Melting Point: 100-102° MDL No.: MFCD01318111

Formel: C₄H₂BrIN₂
Schmelzpunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318111
CAS-Nummer: 183438-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR