4-Isopropylphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Isopropylbenzeneboronic acid
L17459.03
L17459.03EA
98.8
EUR
L17459.03
4-Isopropylphenylboronsäure ≥98%
4-Isopropylphenylboronsäure
Purity: 98+%
CAS No.: 16152-51-5
Molecular Formula: C9H13BO2
Formula Weight: 164.01
Melting Point: 142-144°
Beilstein Reference: 2830262
Formel:
C₉H₁₃BO₂ Molecular Weight: 164,01 g/mol Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01074614 CAS-Nummer: 16152-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...