tert-Butyl pyrrolidine-1-carboxylate 98%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
tert-butyl pyrrolidine-1-carboxylate, pyrrolidine-1-carboxylic acid tert-butyl ester, N-BOC-Pyrrolidine
L17475.06
L17475.06EA
71.4
EUR
L17475.06
tert-Butyl pyrrolidine-1-carboxylate 98%
tert-Butyl pyrrolidine-1-carboxylate
Purity: 98%
CAS No.: 86953-79-9
Molecular Formula: C9H17NO2
Formula Weight: 171.24
Storage Temperature: 2°C to 8°C
Flash Point: 86°(186°F)
Refractive Index: 1.4490
Boiling Point: 130°/20mm
Beilstein Reference: 4664750
MDL No.: MFCD00216581
Formel:
C₉H₁₇NO₂ Molecular Weight: 171,24 g/mol Siedepunkt: 130 °C (20 mmHg) Dichte: 0,977 Flammpunkt: 86 °C (186 °F) |
MDL:
MFCD00216581 CAS-Nummer: 86953-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...