4-(2-Carboxyethyl)phenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-(2-Carboxyethyl)benzeneboronic acid
L17485.03
L17485.06
L17485.03EA
94.2
EUR
L17485.03
L17485.06
4-(2-Carboxyethyl)phenylboronsäure ≥97%
4-(2-Carboxyethyl)phenylboronsäure
Purity: 97%
CAS No.: 166316-48-9
Molecular Formula: C9H11BO4
Formula Weight: 193.99
Melting Point: 146-148°
Beilstein Reference: 9198524
Formel:
C₉H₁₁BO₄ Molecular Weight: 193.99 g/mol Schmelzpunkt: 146…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318119 CAS-Nummer: 166316-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...