6-Methylindole-3-carboxyaldehyde 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Methyl-1H-indole-3-carbaldehyde

L17508.03
L17508.03EA 104 EUR
L17508.03
6-Methylindole-3-carboxyaldehyde 97%
6-Methylindole-3-carboxyaldehyde

Purity: 98% CAS No.: 4771-49-7 Molecular Formula: C10H9NO Formula Weight: 159.19 Melting Point: 185-189° Beilstein Reference: 386914 MDL No.: MFCD00049347

Formel: C₁₀H₉NO
Molecular Weight: 159,19 g/mol
Schmelzpunkt: 185…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00049347
CAS-Nummer: 4771-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR