2-Chlor-5-nitrobenzoylchlorid 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Chlor-5-nitrobenzol-1-carbonylchlorid, 2-Chlor-5-nitrobenzoesäurechlorid

L17523.14
L17523.14EA 52.6 EUR
L17523.14
2-Chlor-5-nitrobenzoylchlorid 96%
2-Chlor-5-nitrobenzoylchlorid

Purity: 96% CAS No.: 25784-91-2 Molecular Formula: C7H3Cl2NO3 Formula Weight: 220.01 Melting Point: 58-60° Boiling Point: 157-158°/11mm UN No.: 3261 MDL No.: MFCD00059180

Formel: C₇H₃Cl₂NO₃
Molecular Weight: 220.01 g/mol
Siedepunkt: 157…158 °C (11 mmHg)
Schmelzpunkt: 58…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00059180
CAS-Nummer: 25784-91-2
EINECS: 247-262-4
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR