1-Boc-piperidin-4-carbonsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid

L17527.03 L17527.06
L17527.03EA 36.8 EUR
L17527.03 L17527.06
1-Boc-piperidin-4-carbonsäure ≥98%
1-Boc-piperidin-4-carbonsäure

Purity: 98+% CAS No.: 84358-13-4 Molecular Formula: C11H19NO4 Formula Weight: 229.28 Melting Point: 149-151° MDL No.: MFCD00076999

Formel: C₁₁H₁₉NO₄
Schmelzpunkt: 149…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00076999
CAS-Nummer: 84358-13-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR