N-(5-Brompentyl)phthalimid ≥97%

Lieferant: Thermo Fisher Scientific

L17543.22 L17543.06 L17543.14
L17543.22EA 376 EUR
L17543.22 L17543.06 L17543.14
N-(5-Brompentyl)phthalimid ≥97%
N-(5-Brompentyl)phthalimid

Purity: 97% CAS No.: 954-81-4 Molecular Formula: C13H14BrNO2 Formula Weight: 296.17 Odour: Odourless Melting Point: 58-62° Beilstein Reference: 177015 MDL No.: MFCD00060522

Formel: C₁₃H₁₄BrNO₂
Schmelzpunkt: 58…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060522
CAS-Nummer: 954-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR