4-Benzyloxyindole-3-carbaldehyde 98%
Lieferant: Thermo Fisher Scientific
L17567.03
L17567.03EA
154
EUR
L17567.03
4-Benzyloxyindole-3-carbaldehyde 98%
4-Benzyloxyindole-3-carbaldehyde
Purity: 98%
CAS No.: 7042-71-9
Molecular Formula: C16H13NO2
Formula Weight: 251.28
Melting Point: 167° dec.
Beilstein Reference: 1537328
MDL No.: MFCD00152002
Formel:
C₁₆H₁₃NO₂ Schmelzpunkt: 167 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00152002 CAS-Nummer: 7042-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...