5-Methyl-3-isoxazolcarbonsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L17577.06
L17577.06EA
68.6
EUR
L17577.06
5-Methyl-3-isoxazolcarbonsäure ≥98%
5-Methylisoxazole-3-carboxylic acid
Purity: 98+%
CAS No.: 3405-77-4
Molecular Formula: C5H5NO3
Formula Weight: 127.10
Melting Point: 171-173°
Beilstein Reference: 114094
Formel:
C₅H₅NO₃ Molecular Weight: 127.1 g/mol Schmelzpunkt: 171…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318162 CAS-Nummer: 3405-77-4 EINECS: 222-289-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...