1-Acetylisatin ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-Acetylindoline-2,3-dione

L17597.03 L17597.06 L17597.14
L17597.03EA 13.1 EUR
L17597.03 L17597.06 L17597.14
1-Acetylisatin ≥97%
1-Acetylisatin

Purity: 97% CAS No.: 574-17-4 Molecular Formula: C10H7NO3 Formula Weight: 189.17 Melting Point: 141-143° Beilstein Reference: 113959

Formel: C₁₀H₇NO₃
Molecular Weight: 189.17 g/mol
Schmelzpunkt: 141…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00158542
CAS-Nummer: 574-17-4
EINECS: 209-368-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR