4-(Ethylthio)benzaldehyd ≥97%

Lieferant: Thermo Fisher Scientific

L17624.03 L17624.06
L17624.03EA 50.6 EUR
L17624.03 L17624.06
4-(Ethylthio)benzaldehyd ≥97%
4-(Ethylthio)benzaldehyde

Purity: 98% CAS No.: 84211-94-9 Molecular Formula: C9H10OS Formula Weight: 166.24 Refractive Index: 1.6240 Boiling Point: 151-153°/15mm MDL No.: MFCD01318145

Formel: C₉H₁₀OS
Molecular Weight: 166.24 g/mol
Siedepunkt: 151…153 °C (15 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD01318145
CAS-Nummer: 84211-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR