7-Chlorindol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L17637.03 L17637.MD
L17637.03EA 120 EUR
L17637.03 L17637.MD
7-Chlorindol ≥98%
7-Chlorindol

Purity: 98% CAS No.: 53924-05-3 Molecular Formula: C8H6ClN Formula Weight: 151.60 Melting Point: 57-58° Beilstein Reference: 114743

Formel: C₈H₆ClN
Schmelzpunkt: 57…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01321301
CAS-Nummer: 53924-05-3
EINECS: 258-865-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR