2-Amino-5-brom-3,4-dimethylpyridin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

L17672.06 L17672.03
L17672.06EA 289 EUR
L17672.06 L17672.03
2-Amino-5-brom-3,4-dimethylpyridin 97%
2-Amino-5-bromo-3,4-dimethylpyridine

Purity: 97% CAS No.: 374537-97-0 Molecular Formula: C7H9BrN2 Formula Weight: 201.07 Melting Point: 133-135° UN No.: 2811 MDL No.: MFCD02093947

Formel: C₇H₉BrN₂
Schmelzpunkt: 133…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093947
CAS-Nummer: 374537-97-0
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR