3-Brom-1,8-naphthalsäureanhydrid ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Bromobenzo[de]isochromene-1,3-dione
L17683.03
L17683.03EA
102
EUR
L17683.03
3-Brom-1,8-naphthalsäureanhydrid ≥97%
3-Brom-1,8-naphthalsäureanhydrid
Purity: 97%
CAS No.: 24050-49-5
Molecular Formula: C12H5BrO3
Formula Weight: 277.08
Melting Point: 244-246°
Beilstein Reference: 180001
Formel:
C₁₂H₅BrO₃ Molecular Weight: 277,08 g/mol Schmelzpunkt: 244…246 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00228162 CAS-Nummer: 24050-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...