1-Tritylimidazol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Clotrimazole Impurity F, 1-Trityl-1H-imidazole

L17694.06
L17694.06EA 71.9 EUR
L17694.06
1-Tritylimidazol ≥98%
1-Tritylimidazol

Purity: 98% CAS No.: 15469-97-3 Molecular Formula: C22H18N2 Formula Weight: 310.40 Odour: Odourless Melting Point: 220-224° Beilstein Reference: 238171 MDL No.: MFCD00229427

Formel: C₂₂H₁₈N₂
Molecular Weight: 310,40 g/mol
Schmelzpunkt: 220…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00229427
CAS-Nummer: 15469-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR