2-Chlor-4-methoxypyridin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

L17697.03
L17697.03EA 167 EUR
L17697.03
2-Chlor-4-methoxypyridin ≥99%
2-Chloro-4-methoxypyridine

Purity: 99% CAS No.: 17228-69-2 Molecular Formula: C6H6ClNO Formula Weight: 143.57 Flash Point: 110°(230°F) Refractive Index: 1.5410 Boiling Point: 110-115°/10mm Beilstein Reference: 1423379 MDL No.: MFCD02093951

Formel: C₆H₆ClNO
Siedepunkt: 110…115 °C (10 mmHg)
Dichte: 1,258
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD02093951
CAS-Nummer: 17228-69-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR