1,2-O-Isopropylidene-ɑ-D-glucurono-6,3-lactone ≥98%
Lieferant: Thermo Fisher Scientific
L17765.09
L17765.18
L17765.09EA
56
EUR
L17765.09
L17765.18
1,2-O-Isopropylidene-ɑ-D-glucurono-6,3-lactone ≥98%
1,2-O-Isopropylidene-alpha-D-glucurono-6,3-lactone
Purity: 98+%
CAS No.: 20513-98-8
Molecular Formula: C9H12O6
Formula Weight: 216.19
Melting Point: 119-121°
Optical Rotation: +52° (c=1 in chloroform)
MDL No.: MFCD00061641
Formel:
C₉H₁₂O₆ Schmelzpunkt: 119…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061641 CAS-Nummer: 20513-98-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...