3,4-Difluorbenzolsulfonylchlorid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3,4-Difluorbenzolsulfochlorid, 3,4-Difluorbenzol-1-sulfonylchlorid, 3,4-Difluorbenzolsulfonsäurechlorid

L17816.03 L17816.06
L17816.03EA 40.3 EUR
L17816.03 L17816.06
3,4-Difluorbenzolsulfonylchlorid ≥97%
3,4-Difluorbenzolsulfonylchlorid

Purity: 97% CAS No.: 145758-05-0 Molecular Formula: C6H3ClF2O2S Formula Weight: 212.60 Refractive Index: 1.5140 Boiling Point: 105°/15mm UN No.: 3265 MDL No.: MFCD00153120

Formel: C₆H₃ClF₂O₂S
Siedepunkt: 105 °C (15 mmHg)
Dichte: 1,586
Lagertemperatur: Raumtemperatur
MDL: MFCD00153120
CAS-Nummer: 145758-05-0
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR