3-Fluor-4-formylphenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Fluoro-4-formylbenzeneboronic acid

L17851.03
L17851.03EA 39.9 EUR
L17851.03
3-Fluor-4-formylphenylboronsäure ≥98%
3-Fluor-4-formylphenylboronsäure

Purity: 98% CAS No.: 248270-25-9 Molecular Formula: C7H6BFO3 Formula Weight: 167.93 Melting Point: ca 240° MDL No.: MFCD02093051

Formel: C₇H₆BFO₃
Molecular Weight: 167.93 g/mol
Schmelzpunkt: ∼ 240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093051
CAS-Nummer: 248270-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR