4-Pyridinboronsäurepinakolester ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Pyridine-4-boronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

L17854.03
L17854.03EA 127 EUR
L17854.03
4-Pyridinboronsäurepinakolester ≥98%
4-Pyridinboronsäurepinakolester

Purity: 98% CAS No.: 181219-01-2 Molecular Formula: C11H16BNO2 Formula Weight: 205.07 Melting Point: 150-152° Beilstein Reference: 7919059

Formel: C₁₁H₁₆BNO₂
Molecular Weight: 205,06 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319051
CAS-Nummer: 181219-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR