2-Nitrophenylboronsäure 96%

Lieferant: Thermo Fisher Scientific
Achtung

L17988.06 L17988.03
L17988.06EA 329 EUR
L17988.06 L17988.03
2-Nitrophenylboronsäure 96%
2-Nitrophenylboronsäure

Purity: 96% CAS No.: 5570-19-4 Molecular Formula: C6H6BNO4 Formula Weight: 166.93 Melting Point: 149-151° Beilstein Reference: 3032097

Formel: C₆H₆BNO₄
Schmelzpunkt: 149…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00161358
CAS-Nummer: 5570-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR