3-(4-Methoxybenzoyl)acrylic acid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-(4-Methoxyphenyl)-4-oxobut-2-enoic acid
L18071.06
L18071.03
L18071.06EA
175
EUR
L18071.06
L18071.03
3-(4-Methoxybenzoyl)acrylic acid 98%
3-(4-Methoxybenzoyl)acrylic acid
Purity: 98%
CAS No.: 5711-41-1
Molecular Formula: C11H10O4
Formula Weight: 206.20
Melting Point: 137-140°
Beilstein Reference: 474529
MDL No.: MFCD00020486
Formel:
C₁₁H₁₀O₄ Schmelzpunkt: 137…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020486 CAS-Nummer: 5711-41-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...