2,4-Bis(trifluormethyl)phenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

L18161.06 L18161.03
L18161.06EA 406 EUR
L18161.06 L18161.03
2,4-Bis(trifluormethyl)phenylboronsäure ≥97%
2,4-Bis(trifluormethyl)phenylboronsäure

Purity: 97% CAS No.: 153254-09-2 Molecular Formula: C8H5BF6O2 Formula Weight: 257.93 MDL No.: MFCD01631349

Formel: C₈H₅BF₆O₂
Molecular Weight: 257.93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631349
CAS-Nummer: 153254-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR