α,α,α,2,4,6-Hexafluortoluol 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,4,6-Trifluorobenzotrifluoride, 1,3,5-Trifluoro-2-(trifluoromethyl)benzene

L18167.06
L18167.06EA 91.5 EUR
L18167.06
α,α,α,2,4,6-Hexafluortoluol 96%
α,α,α,2,4,6-Hexafluortoluol

Purity: 99% CAS No.: 122030-04-0 Molecular Formula: C7H2F6 Formula Weight: 200.08 Flash Point: 22°(71°F) Refractive Index: 1.3850 Boiling Point: 105-107° UN No.: 1993 MDL No.: MFCD00190109

Formel: C₇H₂F₆
Molecular Weight: 200,08 g/mol
Siedepunkt: 105…107 °C
Dichte: 1,475
Flammpunkt: 22 °C (71 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00190109
CAS-Nummer: 122030-04-0
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR