Dimethylaminoacetyl chloride hydrochloride ≥85%, tech.

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-(Dimethylamino)acetyl chloride hydrochloride

L18475.06
L18475.06EA 112 EUR
L18475.06
Dimethylaminoacetyl chloride hydrochloride ≥85%, tech.
Dimethylaminoacetyl chloride hydrochloride

Purity: 85% CAS No.: 60853-81-8 Molecular Formula: (CH3)2NCH2COCl.HCl Formula Weight: 158.03 Beilstein Reference: 3980151 UN No.: UN3261 MDL No.: MFCD02094025

Formel: (CH₃)₂NCH₂COCl·HCl
Molecular Weight: 158,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02094025
CAS-Nummer: 60853-81-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR