4-Bromo-3-fluorophenylboronic acid 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Bromo-3-fluorobenzeneboronic acid

L18514.03 L18514.MD
L18514.03EA 61 EUR
L18514.03 L18514.MD
4-Bromo-3-fluorophenylboronic acid 95%
4-Bromo-3-fluorophenylboronic acid

Purity: 95% CAS No.: 374790-97-3 Molecular Formula: C6H5BBrFO2 Formula Weight: 218.82 Melting Point: 300° MDL No.: MFCD02093066

Formel: C₆H₅BBrFO₂
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093066
CAS-Nummer: 374790-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR