Ethyl (R)-4-chloro-3-hydroxybutyrate 97%, ee 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (R)-Ethyl 4-chloro-3-hydroxybutanoate, (R)-4-Chloro-3-hydroxybutyric Acid Ethyl Ester

L18553.03 L18553.06
L18553.03EA 56.2 EUR
L18553.03 L18553.06
Ethyl (R)-4-chloro-3-hydroxybutyrate 97%, ee 96%
Ethyl (R)-4-chloro-3-hydroxybutyrate

Purity: 97% CAS No.: 90866-33-4 Molecular Formula: C6H11ClO3 Formula Weight: 166.61 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4530 Boiling Point: 93-95°/5mm Optical Rotation: +14° (neat) MDL No.: MFCD00211242

Formel: C₆H₁₁ClO₃
Molecular Weight: 166.60 g/mol
Siedepunkt: 93…95 °C (5 mmHg)
Dichte: 1.190
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00211242
CAS-Nummer: 90866-33-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR