(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine 97%

Lieferant: Thermo Fisher Scientific
Gefahr

L18659.03
L18659.03EA 166 EUR
L18659.03
(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine 97%
(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine

Purity: 97% CAS No.: 153837-28-6 Molecular Formula: C18H30N2 Formula Weight: 274.45 Storage Temperature: 2°C to 8°C Melting Point: 36-40° Beilstein Reference: 6957993 Optical Rotation: -90° (c=1 in methanol) UN No.: UN3259 MDL No.: MFCD06795639

Formel: C₁₈H₃₀N₂
Molecular Weight: 274.45 g/mol
Schmelzpunkt: 36…40 °C
MDL: MFCD06795639
CAS-Nummer: 153837-28-6
ADR: 8 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR