4-Brom-2-fluorbenzylalkohol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4-Brom-2-fluorphenyl)methanol
L18679.06
L18679.06EA
58.5
EUR
L18679.06
4-Brom-2-fluorbenzylalkohol ≥97%
4-Brom-2-fluorbenzylalkohol
Purity: 97%
CAS No.: 188582-62-9
Molecular Formula: C7H6BrFO
Formula Weight: 205.03
Melting Point: 31-33°
Beilstein Reference: 7757667
Formel:
C₇H₆BrFO Molecular Weight: 205,03 g/mol Schmelzpunkt: 31…33 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143265 CAS-Nummer: 188582-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...