D-Epoxone 98%
Lieferant: Thermo Fisher Scientific
L18989.03
L18989.06
L18989.03EA
49.7
EUR
L18989.03
L18989.06
D-Epoxone 98%
D-Epoxone
Purity: 98%
CAS No.: 18422-53-2
Molecular Formula: C12H18O6
Formula Weight: 258.27
Melting Point: 101-103°
Beilstein Reference: 1319073
MDL No.: MFCD00063383
Formel:
C₁₂H₁₈O₆ Schmelzpunkt: 101…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063383 CAS-Nummer: 18422-53-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...