5-Methoxy-3-indolylessigsäure ≥98%
Lieferant: Thermo Fisher Scientific
L19073.03
L19073.MD
L19073.03EA
117
EUR
L19073.03
L19073.MD
5-Methoxy-3-indolylessigsäure ≥98%
Purity: 98+%
CAS No.: 3471-31-6
Molecular Formula: C11H11NO3
Formula Weight: 205.22
Melting Point: 145-148° dec.
Beilstein Reference: 187161
Formel:
C₁₁H₁₁NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005638 CAS-Nummer: 3471-31-6 EINECS: 222-438-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...