6-Methoxyindol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-Methoxy-1H-indol, Indol-6-ylmethylether
L19074.03
L19074.MD
L19074.03EA
124
EUR
L19074.03
L19074.MD
6-Methoxyindol ≥98%
6-Methoxyindol
Purity: 98+%
CAS No.: 3189-13-7
Molecular Formula: C9H9NO
Formula Weight: 147.18
Melting Point: 90-94°
Boiling Point: 105°/0.2mm
MDL No.: MFCD00022780
Formel:
C₉H₉NO Molecular Weight: 147.18 g/mol Siedepunkt: 105 °C (0.2 mmHg) Schmelzpunkt: 90…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022780 CAS-Nummer: 3189-13-7 EINECS: 221-689-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...