1-Butyl-3-methylimidazoliumtetrafluoroborat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Butyl-1-methyl-1H-imidazol-3-ium tetrafluoroborate, 1-n-Butyl-3-methylimidazolium tetrafluoroborate, [BMIM] [BF4]

L19087.09 L19087.18
L19087.09EA 132 EUR
L19087.09 L19087.18
1-Butyl-3-methylimidazoliumtetrafluoroborat ≥98%
1-Butyl-3-methylimidazoliumtetrafluoroborate

Purity: 98+% CAS No.: 174501-65-6 Molecular Formula: C8H15BF4N2 Formula Weight: 226.03 Flash Point: None Refractive Index: 1.4220 MDL No.: MFCD03095449

Formel: C₈H₁₅BF₄N₂
Molecular Weight: 226.02 g/mol
Siedepunkt: ∼200 °C (1013 hPa)
Schmelzpunkt: –71 °C
Dichte: 1.210
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095449
CAS-Nummer: 174501-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR